OccuNomix™ High-Vis Vests

With 1 in. (2.5cm) glossy tape stripes

Manufacturer: Occunomix International Inc LUXXGTMYR

 View more versions of this product

Catalog No. 19-520-370

  Check Availability
Add to Cart

    Specifications

    No. of Pockets 1.0
    Size (English) Chest 34 to 42 in.
    Size (Metric) Chest 86.36 to 106.68cm
    Tape Type 1 in. Glossy silver reflective stripes
    Color Yellow
    Size Regular

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit