OccuNomix™ High-Viz Rain Jacket and Pants
Lightweight and breathable for ease in mobility
Manufacturer: Occunomix International Inc LUXTJRYM
 View more versions of this product
Catalog No. 19-520-360
  Check Availability
Add to Cart
    Specifications
    No. of Pockets 2.0
    Material Fabric
    Size (English) Chest 40 to 42 in.
    Size (Metric) Chest 101.6 to 106.6cm
    Seam Type Welded
    Size Medium
    Color Yellow
    Certifications/Compliance ASTM F1891, F2733
    Construction ANSI Polyester, Polyurethane
  • Description & Specifications
Related Products
Product Content Feedback