Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OccuNomix™ OccuLux™ Economy Mesh Hi-Viz Vests
Click to view available options
Size:
2X-Large
4X-Large to 5X-Large
Small to Medium
X-Large
Color:
Orange
Yellow
Specifications
Color | Yellow |
Size | 4X-Large to 5X-Large |
Size (English) Chest | 56 to 58, 60 to 62 in. |
No. of Pockets | 1 |
Size (Metric) Chest | 142.24 to 147.32, 152.4 to 157.48 cm |
Tape Type | 2 in. Silver reflective stripes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction