OccuNomix™ OccuLux™ Flame Resistant Solid Vest
Manufacturer: Occunomix International Inc LUXSSG/FRY2X
 View more versions of this product
Catalog No. 19-161-771
  Check Availability
Add to Cart
    Specifications
    Arc Flash Rating 5.5 Calories/cm2
    Certifications/Compliance ASTM F1506
    Size (English) Chest 48 to 50 in.
    Size (Metric) Chest 121.92 to 127cm
    Color Yellow
    Construction ANSI Modacrylic
    Material Modacrylic
    Size 2X-Large
  • Description & Specifications
Related Products
Product Content Feedback