OccuNomix™ OccuLux™ Flame Resistant Solid Vest
Manufacturer: Occunomix International Inc LUXSSG/FRY5X
 View more versions of this product
Catalog No. 19-161-774
  Check Availability
Add to Cart
    Specifications
    Arc Flash Rating 5.5 Calories/cm2
    Certifications/Compliance ASTM F1506
    Size (English) Chest 60 to 62 in.
    Size (Metric) Chest 152.4 to 157.48cm
    Color Yellow
    Construction ANSI Modacrylic
    Material Modacrylic
    Size 5X-Large
  • Description & Specifications
Related Products
Product Content Feedback