Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ocean Activity Model Products for Science Education

View the impact of temperature and salt on ocean circulation.

Manufacturer:  Neo/Sci Corporation 301050

Catalog No. S32683

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Detailed teacher and student guides

Description

Description

  • Place different water conditions in each cylinder and watch their interaction
  • Use blue and red food dye to show any combination of cold or hot water and salt or freshwater
  • Rheoscopic fluid provides visual representation of ocean circular patterns
  • L x W x H: 14 x 4.25 x 12 in. (35.6 x 10.8 x 30.5cm)

Specifications

Specifications

View the impact of temperature and salt on ocean circulation
Plastic
10.7cm
Teacher's guide and student guides
35.5cm
Ocean activity model
4.25 in.
14 x 4.25 in. (35.5 x 10.7cm)
14 in.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit