Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oceanography Charts Products for Science Education

Depict ocean floor as well as major seas, lakes, and trenches.

Manufacturer:  Scarfo 0102

Catalog No. S45564


Add to cart

Description

Description

  • Full-color maps suitable for framing
  • Large chart: 29 × 39 in. (73.6 × 99cm)
  • Secondary chart: 18 × 25 in. (45.7 × 63.5cm)

Large chart:

  • Explanation of continental drift theory
  • Cutaway views of continents and ocean basins
  • Table of major bodies of water and trenches dimensions
  • Chemical composition and physical properties of sea waters
  • Explanation of the Beaufort scale
  • Chart depicting major surface currents
  • Ocean tides and how they occur
Secondary chart:
  • Made specifically for student review and testing

Specifications

Specifications

Oceanography Chart
Depict floor of our worlds oceans major seas, lakes and trenches
Large chart 29 x 39 in., secondary chart 18 x 25 in.
Each
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit