Learn More
OCRL Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: S166A-26, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242193V
Catalog No. NBP242193V
Add to cart
Description
Description
OCRL Monoclonal antibody specifically detects OCRL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
OCRL
S166A-26
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
OCRL
Fusion protein amino acids 1-901 (full-length) of human INPP5F. Rat: 93% identity (845/904 amino acids identical). Mouse: 91% identity (824/901 amino acids identical) ~50% identity with INPP5b.
Protein G purified
Golgi Apparatus Markers
Primary
4952
Human, Mouse, Rat
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.1.3.36, INPP5F, LOCR, Lowe oculocerebrorenal syndrome protein, NPHL2, OCRL1inositol polyphosphate 5-phosphatase OCRL-1, oculocerebrorenal syndrome of Lowe, phosphatidylinositol polyphosphate 5-phosphatase
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Approx 50% identity with INPP5b.Detects approx 100kDa. Cross-reacts with INPP5b.