Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCRL Mouse anti-Human, Mouse, Rat, FITC, Clone: S166A-26, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242193F

Catalog No. NBP242193F


Add to cart

Description

Description

OCRL Monoclonal antibody specifically detects OCRL in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

OCRL
S166A-26
Immunocytochemistry/Immunofluorescence
Purified
OCRL
Fusion protein amino acids 1-901 (full-length) of human INPP5F. Rat: 93% identity (845/904 amino acids identical). Mouse: 91% identity (824/901 amino acids identical) ~50% identity with INPP5b.
Protein G purified
Golgi Apparatus Markers
Primary
4952
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
EC 3.1.3.36, INPP5F, LOCR, Lowe oculocerebrorenal syndrome protein, NPHL2, OCRL1inositol polyphosphate 5-phosphatase OCRL-1, oculocerebrorenal syndrome of Lowe, phosphatidylinositol polyphosphate 5-phosphatase
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Approx 50% identity with INPP5b.Detects approx 100kDa. Cross-reacts with INPP5b.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit