Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oct-4B Mouse anti-Human, Mouse, Rat, DyLight™ 405, Clone: OTI7H1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273141V
Description
Oct-4B Monoclonal specifically detects Oct-4B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Oct-4B | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI7H1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
POU5F1 | |
Full length recombinant protein of human Oct-4B (NP_002692) produced in HEK293T cells. | |
0.1 mL | |
5460.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title