Learn More
Oct3/4 Mouse, Clone: 40/Oct-3, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565644
Catalog No. BDB565644
Add to cart
Description
Description
Oct3/4 Monoclonal antibody specifically detects Oct3/4 in Human, Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Specifications
Oct3/4
40/Oct-3
Brilliant Violet 421
Purified
Murine
Mouse IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Oct3, OTF3, Oct4, OTF4, POU5F1
Mouse Oct3 aa. 252-372 Recombinant Protein
50μg
Cell Biology
Primary
Human, Murine
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.