Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT4 Mouse anti-Human, Clone: OCT4/3508, Novus Biologicals™

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP31374420UG

 View more versions of this product

Catalog No. NB177445


Add to Cart
This item is not returnable. View return policy

Description

Description

OCT4 Monoclonal antibody specifically detects OCT4. in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

OCT4.
Monoclonal
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Q01860
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Immunohistochemistry (Paraffin)
OCT4/3508
Unconjugated
10 mM PBS with 0.05% BSA
class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein
Recombinant fragment of human OCT4 protein (around aa 1-134) (exact sequence is proprietary) (Uniprot: Q01860)
20 μg
Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators
5460
Store at 4°C.
IgG2b κ
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit