Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Octacarbonyldicobalt, stab. with 1-5% hexane

As a catalyst in oxo process, and in high-purity cobalt salts

Manufacturer:  Alfa Aesar™ 1306088

Quantity 2.5g

 View more versions of this product

Catalog No. AA1306088


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10210-68-1
341.95
UN3124
Octacarbonyldicobalt, Stabilized with 1 to 5% hexane
Co2(CO)8
MFCD00016024
Cobalt carbonyl; Cobalt tetracarbonyl

Specifications

10210-68-1
Crystalline
341.95
UN3124
Insoluble in water; Soluble in alcohol, ether and carbon disulfide
Red-orange
−13° (−9°F)
Octacarbonyldicobalt, Stabilized with 1 to 5% hexane
1.73g/mL
Co2(CO)8
MFCD00016024
Cobalt carbonyl; Cobalt tetracarbonyl
≈51°C (dec.)
2.5g
Air Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H252-H330-H361-H373-H302-H317
Flammable solid.
Self-heating in large quantities
may catch fire.
Fatal if inhaled.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P210-P304+P340-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, SELF-HEATING, N.O.S.

RTECSNumber : GG0300000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit