Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octacosane, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
630-02-4
C28H58
394.76
MFCD00009355
ZYURHZPYMFLWSH-UHFFFAOYSA-N
n-octacosane,n-octcosane,unii-vff49836p8,ch3-ch2 26-ch3,octacosane, analytical standard,ccris 680,octacosane, n,octacosane,heptacosane, methyl,n-octacosane 10g
12408
CHEBI:32943
octacosane
CCCCCCCCCCCCCCCCCCCCCCCCCCCC
630-02-4
C28H58
394.76
MFCD00009355
ZYURHZPYMFLWSH-UHFFFAOYSA-N
n-octacosane,n-octcosane,unii-vff49836p8,ch3-ch2 26-ch3,octacosane, analytical standard,ccris 680,octacosane, n,octacosane,heptacosane, methyl,n-octacosane 10g
12408
CHEBI:32943
octacosane
CCCCCCCCCCCCCCCCCCCCCCCCCCCC
Color White
Quantity 100g
Melting Point 61.0°C to 64.0°C
Packaging Glass bottle
Specific Gravity 0.8067
Infrared Spectrum Authentic
Boiling Point 440.0°C
Flash Point 227°C
Density 0.8067g/mL
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3(CH2)26CH3
Beilstein 01,I,71
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform
Formula Weight 394.76
Physical Form Shiny Flakes or Powder
Percent Purity ≥99%
Chemical Name or Material Octacosane
Show More Show Less