Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecane, 99+%, ACROS Organics™

Click to view available options
Quantity:
5mL
250mL
1L
Packaging:
Glass bottle
593-45-3
C18H38
254.5
RZJRJXONCZWCBN-UHFFFAOYSA-N
n-octadecane,octadecan,unii-n102p6haiu,n102p6haiu,octadecane,octadecane, analytical standard,octadecane, n,oktadekan,ccris 681,stearyl group
11635
CHEBI:32926
octadecane
CCCCCCCCCCCCCCCCCC
593-45-3
C18H38
254.5
RZJRJXONCZWCBN-UHFFFAOYSA-N
n-octadecane,octadecan,unii-n102p6haiu,n102p6haiu,octadecane,octadecane, analytical standard,octadecane, n,oktadekan,ccris 681,stearyl group
11635
CHEBI:32926
octadecane
CCCCCCCCCCCCCCCCCC
Quantity 5mL
Melting Point 28.0°C to 31.0°C
Packaging Glass bottle
Boiling Point 317.0°C
Color White
Density 0.7770g/mL
Flash Point 166°C
Infrared Spectrum Authentic
Specific Gravity 0.777
Assay Percent Range 99+%
Linear Formula CH3(CH2)16CH3
Beilstein 01,173
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol, acetone and diethyl ether
Formula Weight 254.5
Physical Form Crystalline Low Melting Solid
Percent Purity ≥99%
Chemical Name or Material Octadecane
Show More Show Less

GHS P Statement May be fatal if swallowed and enters airways. Repeated exposure may cause skin dryness or cracking.

GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting.

Warning