Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecylamine, 90%, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
124-30-1
C18H39N
269.51
MFCD00008159
REYJJPSVUYRZGE-UHFFFAOYSA-N
octadecylamine,1-octadecanamine,stearylamine,1-octadecylamine,octadecanamine,stearamine,1-aminooctadecane,n-octadecylamine,n-stearylamine,armofilm
15793
CHEBI:63866
octadecan-1-amine
CCCCCCCCCCCCCCCCCCN
124-30-1
C18H39N
269.51
MFCD00008159
REYJJPSVUYRZGE-UHFFFAOYSA-N
octadecylamine,1-octadecanamine,stearylamine,1-octadecylamine,octadecanamine,stearamine,1-aminooctadecane,n-octadecylamine,n-stearylamine,armofilm
15793
CHEBI:63866
octadecan-1-amine
CCCCCCCCCCCCCCCCCCN
Viscosity 4.17 mPa.s (20°C)
Water 0.5% max.
Packaging Plastic drum
Boiling Point 232.0°C (32.0 mmHg)
Infrared Spectrum Authentic
Quantity 5kg
Flash Point 148°C
Melting Point 50.0°C to 60.0°C
Color White
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula CH3(CH2)17NH2
Beilstein 04, IV, 825
Concentration or Composition (by Analyte or Components) Primary amine (MW 269):> 98%, Sec. + tert. amine (MW 521): < 2%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons
Formula Weight 269.51
Physical Form Prills
Percent Purity 90%
Grade Technical
Chemical Name or Material Octadecylamine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger