Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecylbenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
4445-07-2
C24H42
330.59
MFCD00048500
WSVDSBZMYJJMSB-UHFFFAOYSA-N
1-phenyloctadecane,benzene, octadecyl,n-octadecylbenzene,stearylbenzene,2-phenyloctadecane,octadecane, 1-phenyl,n-octadecyl benzene,acmc-2097fm,octadecane, 1-phenyl-8ci,1-phenyloctadecane, analytical standard
78187
octadecylbenzene
CCCCCCCCCCCCCCCCCCC1=CC=CC=C1
4445-07-2
C24H42
330.59
MFCD00048500
WSVDSBZMYJJMSB-UHFFFAOYSA-N
1-phenyloctadecane,benzene, octadecyl,n-octadecylbenzene,stearylbenzene,2-phenyloctadecane,octadecane, 1-phenyl,n-octadecyl benzene,acmc-2097fm,octadecane, 1-phenyl-8ci,1-phenyloctadecane, analytical standard
78187
octadecylbenzene
CCCCCCCCCCCCCCCCCCC1=CC=CC=C1
Boiling Point 249°C (15.0mmHg)
Flash Point >112°C
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Yellow
Melting Point 32°C to 36°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5(CH2)17CH3
Formula Weight 330.59
Physical Form Low Melting Solid
Percent Purity ≥97.5%
Chemical Name or Material Octadecylbenzene
Show More Show Less