Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecylbenzene, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
4445-07-2
C24H42
330.60
MFCD00048500
WSVDSBZMYJJMSB-UHFFFAOYSA-N
1-naphthalen-2-yl prop-2-en-1-one,1-2-naphthyl prop-2-en-1-one,jak3 inhibitor v,2-naphthylvinyl ketone,2'-acrylonaphthone,2-acryloylnaphthalene,thr1 inhibitor, 1,2-naphthyl vinyl ketone,biomolki_000064,biomolki2_000005
3799
octadecylbenzene
CCCCCCCCCCCCCCCCCCC1=CC=CC=C1
4445-07-2
C24H42
330.60
MFCD00048500
WSVDSBZMYJJMSB-UHFFFAOYSA-N
1-naphthalen-2-yl prop-2-en-1-one,1-2-naphthyl prop-2-en-1-one,jak3 inhibitor v,2-naphthylvinyl ketone,2'-acrylonaphthone,2-acryloylnaphthalene,thr1 inhibitor, 1,2-naphthyl vinyl ketone,biomolki_000064,biomolki2_000005
3799
octadecylbenzene
CCCCCCCCCCCCCCCCCCC1=CC=CC=C1
Boiling Point 249°C (15.0mmHg)
Flash Point >112°C
Melting Point 32°C to 36°C
Color White to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 50g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5(CH2)17CH3
Formula Weight 330.59
Physical Form Low Melting Solid
Percent Purity ≥97.5%
Chemical Name or Material Octadecylbenzene
Show More Show Less