Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Octafluorotoluene, 98+%

Manufacturer:  Alfa Aesar™ A1145306

Quantity 5g

 View more versions of this product

Catalog No. AAA1145306


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

434-64-0
236.06
UN1992
Perfluorotoluene
C7F8
MFCD00000375
2057072
Octafluorotoluene

Specifications

434-64-0
236.06
UN1992
Perfluorotoluene
−66°C
20°C (68°F)
≥98%
C7F8
MFCD00000375
2057072
103° to 105°C
5g
Octafluorotoluene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H301-H331-H315-H319-H335
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : XT3530000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit