Alfa Aesar™ Octafluorotoluene, 98+%

Manufacturer: Alfa Aesar™ A1145322

Quantity 100g

 View more versions of this product

Catalog No. AAA1145322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 434-64-0
    Molecular Formula C7F8
    Formula Weight 236.06
    MDL Number MFCD00000375
    UN Number UN1992
    Beilstein 2057072
    Synonym Perfluorotoluene
    Chemical Name or Material Octafluorotoluene

    Specifications

    Formula Weight 236.06
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7F8
    CAS 434-64-0
    Quantity 100g
    TSCA TSCA
    Beilstein 2057072
    Health Hazard 1 Danger
    Melting Point −66°C
    Health Hazard 2 GHS H Statement
    H225-H301-H331-H315-H319-H335
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    RTECS Number XT3530000
    MDL Number MFCD00000375
    UN Number UN1992
    Synonym Perfluorotoluene
    Boiling Point 103° to 105°C
    Flash Point 20°C (68°F)
    Chemical Name or Material Octafluorotoluene
    Assay ≥98%

    GHS H Statement
    H225-H301-H331-H315-H319-H335
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit