Alfa Aesar™ Octanenitrile, 97%

Manufacturer: Alfa Aesar™ B2333814

Quantity 25g

 View more versions of this product

Catalog No. AAB2333814

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 124-12-9
    Molecular Formula C8H15N
    Linear Formula CH3(CH2)6CN
    Formula Weight 125.21
    MDL Number MFCD00001983
    Synonym 1-Cyanoheptane; n-Heptyl cyanide
    Chemical Name or Material Octanenitrile

    Specifications

    Formula Weight 125.21
    Linear Formula CH3(CH2)6CN
    MDL Number MFCD00001983
    Synonym 1-Cyanoheptane; n-Heptyl cyanide
    Health Hazard 1 Warning
    Boiling Point 198° to 200°C
    Odor Characteristic
    Health Hazard 3 GHS P Statement
    P280f
    Wear protective clothing.
    TSCA TSCA
    Molecular Formula C8H15N
    CAS 124-12-9
    Melting Point −45°C
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H302-H227
    Harmful if swallowed.
    Combustible liquid.
    Chemical Name or Material Octanenitrile
    Assay 97%

    GHS H Statement
    H302-H227
    Harmful if swallowed.
    Combustible liquid.

    GHS P Statement
    P280f
    Wear protective clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit