Learn More
Octanoic acid, 98+%, Thermo Scientific Chemicals
Description
Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.
Solubility
Misicible with alcohol, chloroform, ether, carbon disulfide, petroleum ether and acetonitrile.Slightly miscible with water.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications
Melting Point | 16°C to 17°C |
Density | 0.91 |
pH | 4 |
Boiling Point | 236°C to 238°C |
Flash Point | 132°C (269°F) |
Odor | Rancid |
Assay Percent Range | >98% |
Refractive Index | 1.4275 |
Linear Formula | CH3(CH2)6CO2H |
Quantity | 25 mL |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 204-677-5 |
RTECSNumber | RH0175000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.