Alfa Aesar™ Octenyltrichlorosilane, tech.

Manufacturer: Alfa Aesar™ H5343406

Quantity 5g

 View more versions of this product

Catalog No. AAH5343406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 153447-97-3
    Molecular Formula C8H15Cl3Si
    Linear Formula CH2=CH(CH2)6SiCl3
    Formula Weight 245.65
    MDL Number MFCD00039292
    UN Number UN2987
    Chemical Name or Material Octenyltrichlorosilane
    Grade Technical Grade

    Specifications

    Formula Weight 245.65
    Molecular Formula C8H15Cl3Si
    Linear Formula CH2=CH(CH2)6SiCl3
    MDL Number MFCD00039292
    UN Number UN2987
    Boiling Point 223°C
    Quantity 5g
    Flash Point 94°C (201°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
    CAS 153447-97-3
    Chemical Name or Material Octenyltrichlorosilane
    Grade Technical Grade

    Description

    Applications
    Multiwalled carbon nanotubes functionalized with 7-octenyltrichlorosilane and n-octyltrichlorosilane. A novel fabrication method of organic multilayer film is by using 7-octenyltrichlorosilane. Octenyltrichlorosilane is used as functionalizing agent of the raw, purified and functionalized nanotubes.

    Solubility
    Reacts with water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit