OctMAB, Tocris™

Dynamin inhibitor

Manufacturer: R&D Systems 4225/50

Quantity 50mg

 View more versions of this product

Catalog No. 42-255-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C21H46BrN
    Formula Weight 392.5g/mol
    Synonym Octadecyltrimethylammonium bromide
    Chemical Name or Material OctMAB

    Specifications

    Quantity 50mg
    CAS 1120-02-1
    Assay Percent Range >98%
    Molecular Formula C21H46BrN
    Formula Weight 392.5g/mol
    Synonym Octadecyltrimethylammonium bromide
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material OctMAB

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit