Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octyl Phenol Ethoxylate, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number 02003561
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Flash Point 251.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 120 mL
Specific Gravity 1L = 1.07kg
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Octyl Phenol Ethoxylate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit