Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octyltrichlorosilane, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
5283-66-9
C8H17Cl3Si
247.67
MFCD00000488
RCHUVCPBWWSUMC-UHFFFAOYSA-N
trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
21354
trichloro(octyl)silane
CCCCCCCC[Si](Cl)(Cl)Cl
5283-66-9
C8H17Cl3Si
247.67
MFCD00000488
RCHUVCPBWWSUMC-UHFFFAOYSA-N
trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
21354
trichloro(octyl)silane
CCCCCCCC[Si](Cl)(Cl)Cl
Boiling Point 233°C (731.0mmHg)
Viscosity 1.9 mPa.s (25°C)
Flash Point 96°C
Specific Gravity 1.07
Refractive Index 1.4470 to 1.449
Density 1.0700g/mL
Color Colorless to Yellow
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3(CH2)7SiCl3
Beilstein 04, III, 1907
Solubility Information Solubility in water: reacts
Formula Weight 247.67
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Octyltrichlorosilane
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 226-112-1