Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ocular development associated gene Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: OTI1H1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273142UV
Description
Ocular development associated gene Monoclonal specifically detects Ocular development associated gene in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Ocular development associated gene | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GATAD1 | |
Recombinant protein fragment corresponding to amino acids 68-269 of human Ocular development associated gene (NP_066990) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI1H1 | |
Western Blot | |
FLJ22489, GATA zinc finger domain containing 1, GATA zinc finger domain-containing protein 1, ocular development associated, Ocular development-associated gene protein, ODAGFLJ40695, RG083M05.2 | |
Mouse | |
Affinity Purified | |
Vision | |
57798.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title