Learn More
ODC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 170031AP
Catalog No. 50-173-1322
Add to cart
Description
Description
Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis.
Specifications
Specifications
ODC1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P00860, P09057, P11926
ODC, ODC1, Ornithine decarboxylase, ornithine decarboxylase 1
Rabbit
IgG
150 μL
-20°C
Primary
18263, 24609, 4953
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ODC1
Liquid
Odc1
ODC1 Fusion Protein Ag10699
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat