Learn More
Aviva Systems Biology Corp ODF2L Antibody - middle region (ARP56176_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56176_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ODF2L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ODF2L. Alternate names: KIAA1229, MGC111060, RP5-977L11.1, dJ977L11.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-61749