Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODF2L Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548495

Catalog No. PA548495

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine based on sequence homology.

Specifications

Specifications

ODF2L
Polyclonal
PBS with 0.09% sodium azide
Q9ULJ1
Outer dense fiber protein 2-like, ODF2L, KIAA1229
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57489
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ODF2L
Liquid
ODF2L
KLH conjugated synthetic peptide between 543-572 amino acids from the C-terminal region of human ODF2L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit