Learn More
ODF3 Rabbit anti-Human, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 250971AP
Catalog No. 50-173-4721
Add to cart
Description
Description
Outer dense fibers are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail. May help to maintain the passive elastic structures and elastic recoil of the sperm tail.
Specifications
Specifications
ODF3
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96PU9
hSHIPPO, ODF3, Outer dense fiber protein 3, SHIPPO1, Sperm tail protein SHIPPO 1, TISP50
Rabbit
IgG
150 μL
-20°C
Primary
113746, 365387
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ODF3
Liquid
Odf3
ODF3 Fusion Protein Ag18734
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat