Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ODZ3 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281974MFV500
Description
ODZ3 Polyclonal antibody specifically detects ODZ3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| ODZ3 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| FLJ10474, FLJ10886, KIAA1455odd Oz/Ten-m homolog 3, odz, odd Oz/ten-m homolog 3 (Drosophila), ODZ3-like protein, Protein Odd Oz/ten-m homolog 3, ten-3, tenascin-M3, teneurin-3, Ten-m3, TNM3 | |
| ODZ3 antibody was raised against a 17 amino acid peptide near the amino terminus of human ODZ3. The immunogen is located within the first 50 amino acids of ODZ3. Amino Acid Squence: SLTKSRREKERRYTNS | |
| 0.1 mL | |
| Vision | |
| 55714 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction