Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ Offset High Vacuum Hollow Plug Stopcock

Stem OD: 8, 10 or 12 mm available, with hollow glass plug, Precision ground with corresponding numbers on the barrel and the plug.
Supplier: Synthware™ S701754
Specifications
Offset Glass High Vacuum Valve | |
Borosilicate Glass | |
Organic Synthesis |
4 mm | |
10 mm | |
Glass |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title