Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ofloxacin, 98%

Catalog Number p-7077572
Click to view available options
Quantity:
1 g
5 g
25 g
82419-36-1
C18H20FN3O4
361.373
GSDSWSVVBLHKDQ-UHFFFAOYSA-N
4583
CHEBI:7731
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O
82419-36-1
C18H20FN3O4
361.373
GSDSWSVVBLHKDQ-UHFFFAOYSA-N
4583
CHEBI:7731
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O

Ofloxacin, CAS # 82419-36-1, is a fluoroquinolone antibiotic with broad antibacterial activity against gram-positive and -negative bacteria, as well as against mycobacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
Ofloxacin is a synthetic fluoroquinolone antibiotic that is an analog of nalidixic acid
Ofloxacin can directly interact with pneumococcal autolysin (LytA) at its choline binding domain, thus inhibiting the ability of LytA to degrade the pneumococcal cell wall
Ofloxacin is racemic and its (S)-enantiomer is levofloxacin
Applications
Ofloxacin may be used in pharmacokinetic laboratory studies and high-throughput drug screens
The DNA and protein-binding capabilities of ofloxacin have been studied in the laboratory
TRUSTED_SUSTAINABILITY
Type Ofloxacin
Melting Point 280.0°C to 275.0°C
Infrared Spectrum Authentic
Loss on Drying 0.2% max.
Solubility Information Solubility in water: slightly soluble.
Formula Weight 361.37
Percent Purity 98%
Quantity 1 g
Chemical Name or Material Ofloxacin

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit