Learn More
Invitrogen™ OGDH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117043
Description
Positive Samples: Mouse kidney, Mouse heart, Mouse brain, Rat kidney Immunogen sequence: DPLGILDADL DSSVPADIIS STDKLGFYGL DESDLDKVFH LPTTTFIGGQ ESALPLREII RRLEMAYCQH IGVEFMFIND LEQCQWIRQK FETPGIMQFT NEEKRTLLAR LVRSTRFEEF LQRKWSSEKR FGLEGCEVLI PALKTIIDKS SENGVDYVIM GMPHRGRLNV LANVIRKELE QIFCQFDSKL EAADEGSGDV KYHLGMYHRR INRVTDRNIT LSLVANPSHL EAADPVVMGK TKAEQFYCGD TEGKKVRPRE RRARQIVKAP CSSMEFRSPT.
This gene encodes one subunit of the 2-oxoglutarate dehydrogenase complex. This complex catalyzes the overall conversion of 2-oxoglutarate (alpha-ketoglutarate) to succinyl-CoA and CO(2) during the Krebs cycle. The protein is located in the mitocondrial matrix and uses thiamine pyrophosphate as a cofactor. A congential deficiency in 2-oxoglutarate dehydrogenase activity is believed to lead to hypotonia, metabolic acidosis, and hyperlactatemia.
Specifications
OGDH | |
Polyclonal | |
Unconjugated | |
OGDH | |
2210403E04Rik; 2210412K19Rik; 2-oxoglutarate dehydrogenase complex component E1; 2-oxoglutarate dehydrogenase E1 component, mitochondrial; 2-oxoglutarate dehydrogenase, mitochondrial; AA409584; AKGDH; Alpha-ketoglutarate dehydrogenase; d1401; E1k; Kiaa4192; mKIAA4192; OGDC; OGDC-E1; Ogdh; Ogdhl; oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide); oxoglutarate decarboxylase; oxoglutarate dehydrogenase; oxoglutarate dehydrogenase (lipoamide); oxoglutarate dehydrogenase (succinyl-transferring); testicular tissue protein Li 131 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18293, 360975, 4967 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q02218, Q5XI78, Q60597 | |
OGDH | |
Recombinant fusion protein containing a sequence corresponding to amino acids 148-427 of human OGDH (NP_0010039411). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.