Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OGFOD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531548
Description
Recommended positive controls: mouse brain, rat brain. Predicted reactivity: Mouse (87%), Rat (86%), Pig (92%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
OGFOD1, a member of the 2-OG-Fe(II) dioxygenase family, plays an important role in ischemic survival and stress. It is an oxidoreductase containing a Prolyl 4-hydroxylase alpha subunit. An OGFOD1 iron binding residue is required for ATPAF1 gene expression. As a novel stress granule component, OGFOD1 plays important proapoptotic roles in the regulation of translation and HRI-mediated phosphorylation of eIF2alpha in cells.
Specifications
OGFOD1 | |
Polyclonal | |
Unconjugated | |
OGFOD1 | |
2-oxoglutarate and iron dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1; 4930415J21Rik; AA387199; AA939912; AW061076; Kiaa1612; mKIAA1612; OGFOD1; Prolyl 3-hydroxylase OGFOD1; RGD1308848; Termination and polyadenylation 1 homolog; TPA1; TPA1, termination and polyadenylation 1, homolog; uS12 prolyl 3-hydroxylase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
270086, 307657, 55239 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q3U0K8, Q8N543 | |
OGFOD1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 255 of Human OGFOD1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction