Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OGFOD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP276391

Catalog No. NBP276391


Add to cart

Description

Description

OGFOD1 Polyclonal antibody specifically detects OGFOD1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

OGFOD1
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
2-oxoglutarate and iron-dependent oxygenase domain containing 1,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1, EC 1.14.11, FLJ10826, KIAA1612TPA1, termination and polyadenylation 1, homolog, Termination and polyadenylation 1 homolog, TPA1EC 1.14.11.-
Rabbit
IgG
100 μL
Primary
55239.0
Immunoprecipitation, Western Blot
Western Blot 1:1000, Immunoprecipitation 20-40 μL/mg lysate
Purified
OGFOD1
Immunogen sequence between amino acids 1 and 50
Antigen Affinity-purified
Store at 2-8°C/ 1 year from date of receipt
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit