Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OGFOD1 Mouse anti-Human, Clone: OTI1D9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525473

Catalog No. PIMA525473


Add to cart

Description

Description

OGFOD1, a member of the 2-OG-Fe(II) dioxygenase family, plays an important role in ischemic survival and stress. It is an oxidoreductase containing a Prolyl 4-hydroxylase alpha subunit. An OGFOD1 iron binding residue is required for ATPAF1 gene expression. As a novel stress granule component, OGFOD1 plays important proapoptotic roles in the regulation of translation and HRI-mediated phosphorylation of eIF2alpha in cells.
Specifications

Specifications

OGFOD1
Monoclonal
0.41 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N543
TPA1; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1; TPA1, termination and polyadenylation 1, homolog; termination and polyadenylation 1 homolog
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55239
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1D9
Unconjugated
OGFOD1
Liquid
OGFOD1
Full length human recombit protein of OGFOD1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit