Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OGFOD2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176533MFV450
Description
OGFOD2 Polyclonal antibody specifically detects OGFOD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
OGFOD2 | |
Polyclonal | |
Western Blot, ELISA | |
2-oxoglutarate and iron-dependent oxygenase domain containing 2, DKFZp686H15154, EC 1.14.11.-, FLJ13491, FLJ37501, MGC120434, MGC120436,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2 | |
Antibody was raised against a 17 amino acid synthetic peptide near the center of human OGFOD2. The immunogen is located within amino acids 140 - 190 of OGFOD2. Amino Acid Squence: EHFEQSDMPKGRPNTMN | |
0.1 mL | |
Apoptosis | |
79676 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction