Learn More
Bachem Octyl glucoside
Supplier:  Bachem 4005687.0001
Encompass_Preferred
Octyl glucoside, 29836-26-8, C14H28O6, Mr 292.37, 1g. OG, n-Octyl-b-D-glucopyranoside, OGP. OGP, non-ionic, dialyzable detergent for the solubilization and isolation of membrane proteins.
Catalog No. 50-194-7405