Learn More
Tocris Bioscience™ OGT 2115
Antiangiogenic. Heparanase inhibitor
$103.90 - $220.50
Specifications
Chemical Name or Material | 2-[4-[[3-(4-Bromophenyl)-1-oxo-2-propenyl]amino]-3-fluorophenyl]-5-benzoxazoleacetic acid |
---|---|
CAS | 853929-59-6 |
Assay Percent Range | >98% |
Solubility | Soluble to 10mM in DMSO |
Molecular Formula | C24H16BrFN2O4 |
Description
Heparanase inhibitor (IC50 = 0.4 μM) that displays no major inhibition of human cytochrome P450 isoenzymes (IC50 > 30 μM). Exhibits antiangiogenic properties in vitro (IC50 = 1 μM).Specifications
2-[4-[[3-(4-Bromophenyl)-1-oxo-2-propenyl]amino]-3-fluorophenyl]-5-benzoxazoleacetic acid | |
>98% | |
C24H16BrFN2O4 | |
97% |
853929-59-6 | |
Soluble to 10mM in DMSO | |
Observed MW: 495.3 |
Safety and Handling
Recommended Storage : Store at Room Temperature
For Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.