Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OHAUS™ 1050mL Bowl For CS Series Compact Scales
For use with CS Series Compact Scales.
Supplier: OHAUS™ 80850075/EMD
Description
- For use with Compact Scale models: CS200 AM, CS2000 AM, CS5000 AM, CS200P, CS2000P, CS5000P
Specifications
CS Series Compact Scale | |
1050 mL | |
Bowl |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction