Learn More
OHAUS™ Adventurer™ Pro Analytical Balances
Rich in features, compact in size
Manufacturer:  OHAUS™ 80103781
 View more versions of this product
Catalog No. 01-919-208
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Analytical Balance
0.1mg
8.6 in.
InCal™ internal calibration, Glass square frameless flip-top draft shield with removable doors
11.87 in.
30cm
50 to 86°F
100/120V
4.4kg
100/120V 50/60Hz
Milligram, gram, kilogram, ounce, pound, carat, pennyweight, ounce troy, grain, Newton, Hong Kong tael, Singapore tael, Taiwan tael, momme, tical, baht, mesghal, tola, custom
260g
12 in.
±0.3mg
<±0.1mg
22cm
CE, FCC, cULus, CCSAUS, NRTL, C Tick, CB Scheme, IP33, IP20
Backlit LCD
50/60Hz
22cm
ABS Plastic, Stainless Steel, Glass
3 sec.
10 to 30°C
9.6 lb.
9cm
30 x 22 x 30.5cm (11.87 x 8.7 x 12 in.)
2 Years
RS232 with GLP/GMP Data Output
30.5cm
AV264C
8.7 in.
Accessories