Learn More
OHAUS™ Adventurer™ Pro Precision Electronic Balances
Simple operation for basic weighing, with the accuracy and range to support more advanced applications
Manufacturer:  OHAUS™ 80103898
 View more versions of this product
Catalog No. 01-919-214
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Precision Balance
CE, UL, CSA, FCC, NRTL, OIML F1 or ASTM Class 2
Not Applicable
3.3 in.
50/60Hz
11.8 in.
30cm
±0.01g
ABS Plastic, Stainless Steel, Glass
50 to 86°F
100/240V
5.7 lb.
19.3cm
30 x 19.3 x 8.5cm
Milligram, gram, kilogram, ounce, pound, carat, pennyweight, ounce troy, grain, Newton, Hong Kong tael, Singapore tael, Taiwan tael, momme, tical, baht, mesghal, tola, custom
2 Years
LCD
RS232 with GLP/GMP data output
4 Button keypad with dedicated yes, no, back and exit functions
0.01g
8.5cm
AC adapter, RS232 interface, In-use cover, Integral security bracket, Weigh below hook, InCal Internal Calibration
Automatic
±0.02g
AV212C w/ Internal Calib.
1.5 sec.
10 to 30°C
2.6kg
7.6 in.
Not Applicable
100/240V 50/60Hz
30 x 22 x 8.5cm
210g
Accessories