Learn More
OHAUS™ CL Portable Balances
Perform basic weighing with a simple two-button keypad. The CL Portable balance is designed with a large weighing platform and can adequately handle large samples including oversized letters.
Manufacturer:  OHAUS™ 80010610
 View more versions of this product
Catalog No. 01-919-234
Due to product restrictions, please to purchase or view availability for this product.
Includes: 3 AA Batteries (Included); AC Adapter (not included)
Description
Description
The CL Portable balance is designed with a large weighing platform and can adequately handle large samples including oversized letters.
  • ABS housing, ABS pan
  • Low battery indicator
  • Auto shut-off feature
  • Mechanical and software overload protection
  • External push-button calibration
  • Liquid Crystal Display (LCD)
  • RF protection for use near cell phones and production areas
  • Rugged housing with integral platform ring for stacking and storage
Specifications
Specifications
200 g
OIML M1 or ASTM CL F
20cm
0.1 g
4cm
±0.1g
CL201
3 sec.
18 to 25°C
0.87 lb.
14cm
Gram, pound, ounce, pennyweight
LCD
Compact Scale
2 Button keypad
7.9 in.
1.6 in.
3 AA Batteries (Included); AC Adapter (Not include
0.1g
ABS Plastic
65 to 77°F
0.4kg
5.4 in.
12cm
1 Year