Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Defender 3000 Bench Scales

Economical and versatile scale weighs up to 600 lb. (300kg) with 1:6000-7500d resolution.

Manufacturer:  OHAUS™ 83998116

 View more versions of this product

Catalog No. 01-919-804


Due to product restrictions, please to purchase or view availability for this product.


Includes: AC adapter, RS-232 cable, painted Carbon Steel frame, tubular painted Steel column, externally visible level bubble and adjustable Rubber leveling feet with locking nuts

Description

Description

  • 304 stainless steel pan with painted carbon steel frame and aluminum IP67 load cell
  • Adjustable nonslip rubber leveling feet with external level indicator
  • T31P with ABS housing or T31XW with IP66 stainless-steel housing
  • Multiple weighing units include lb., oz., lb:oz, kg, g
  • Parts counting mode with selectable sample sizes, last sample size and Average Piece Weight data
  • One 25mm-high LCD weight display with high-contrast backlight
  • Built-in RS232 for printing and data connection
  • AC adapter and internal rechargeable lead acid battery

Service Plan

Service agreement available.

Specifications

Specifications

Defender™ 3000 bench scale
Internal rechargeable Lead Acid battery
ISO 9001:2008, EMC: EN60950, EN61326, FCC Class A, CE, cULus
25.59 in.
LCD
14.7cm
ABS Plastic, Stainless Steel, Aluminum
0.05kg
14 to 104°F
71 lb.
19.68 in.
600 lb., 272 kg
100 hr.
Span 5kg or 5 lb. to 100% of capacity, user selectable
Bi-directional RS232C
65cm
5.8 in.
AC adapter, painted Carbon Steel frame, 27.36 in. tubular painted Steel column, externally visible level bubble and adjustable Rubber leveling feet with locking nuts
D31P300BX
<2 sec.
-10 to 40°C
32kg
50cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit