Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Defender 5000 Bench Scales with Square Base

Heavy-duty scale for industrial processing and distribution applications.

Manufacturer:  OHAUS™ 80501157

 View more versions of this product

Catalog No. 02-113-41


Due to product restrictions, please to purchase or view availability for this product.


Includes: Universal switching (100-240 VAC 5/60Hz) power supply

Requires : Six alkaline “C” batteries (rechargeable NiMH batteries optional).

Description

Description

Large pan provides ample room for large and unusually shaped samples or packages. Operates on batteries or with internal power supply; optional NiMH internal rechargeable battery is available.
 
  • Easy to clean
  • Painted steel frame and stainless-steel platforms
  • Table/wall mount bracket, fully adjustable feet, plug-in load cell connector
  • Impact-resistant plastic housing protects indicator
  • Large, 1 in. (2.5cm) backlit LCD display and three checkweighing LEDs
  • Five user-selectable weighing units (kg, g, lb, oz, lb:oz), custom, and user-defined unit
  • Simple four-button operation makes weighing effortless
  • Overload protection
  • Auto zero tracking to minimize drift
  • Auto-off timer eliminates battery drain (when batteries are used)
  • Adjustable averaging and stability levels
  • RS-232-C bidirectional interface
  • GMP output with time/date

Service Plan

Service agreement available.

Certifications

CE marked, UL listed, CSA listed.

Specifications

Specifications

4 Button membrane switch
Span 1kg or 1 lb. to 100% of capacity/3-point linearity (0 to 50% to 100% fixed)
RS232C bidirectional, external input
45.7cm
100/240V 50/60Hz
4.5 in.
50/60Hz
Universal switching (100-240 VAC 5/60Hz) power supply
D51P100QL2
<2 sec.
-10 to 40°C
66.6 lb.
18 in.
100 kg
6 C Alkaline
ISO 9001:2008, NTEP, Directive 2002/96/EC, c ULus 2005/95/EC, EN60950-1:2001, FCC Part 15 Class A 2004/108/EC EMC directive: EN61326-1:1997, +A1:1998, +A2:2001, + A3>2003
18 in.
LCD
Gram, kilogram, ounce, pound, metric tones, custom
11.4cm
67
ABS Plastic, Stainless Steel, Polyurethane, Aluminum
0.01kg
14 to 104°F
100/240V
30.2kg
45.7cm
Defender™ 5000 bench scale
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit