Learn More
OHAUS™ Defender™ 7000 Bench Scales
Offers multiple features, durability, and easy configuration. Ideal for shipping and receiving, production, packaging and general commercial and industrial weighing applications.
Manufacturer:  OHAUS™ 80501477
 View more versions of this product
Catalog No. 01-919-805
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  •  Full alphanumeric keypad, dual-line display supporting English, Spanish or French text, checkweighing LEDs
  • Advanced parts counting with dual-scale counting, dynamic weighing, percent weighing, display hold, checkweighing, accumulation, time/date
  • Temporary display of 10x readability in weighing mode using quick key press
  • 255 item library data storage function
  • GMP output with date time, statistics, lb., oz., lb.:oz., g, kg, metric tonnes, user-defined custom unit
  • 304 grade stainless-steel platform pan with black polyurethane painted steel form-and weld frame, IP67 aluminium load cell, stainless-steel column and wall bracket included
  • LCD display with white LED backlight, alphanumeric line and multi-language, plus 3 color checkweigh LED
  • RS232 interface with GMP, Time-Date, optional RS422/485, AC/DC Relay, 2nd RS232 interface
  • Internal universal power supply, 6 x C batteries or optional internal NiMH rechargeable battery
Specifications
Specifications
0.0001kg
14 to 104°F
100/240V
14.7kg
12 in.
30.5cm
Defender™ 7000 bench scale
80 hr.
Span 1kg or 1 lb. to 100% of capacity/linearity: 3 or 5 point
RS232, external input
12 in.
30.5cm
100/240V 50/60Hz
3.7 in.
50/60Hz
Painted Steel column
D71P10QR1
<2 sec.
-10 to 40°C
32.4 lb.
12 in.
30.5cm
25 lb., 10 kg
Positive feedback membrane keys, 5 function keys and alpha-numeric keypad
6 C
ISO 9001:2008, NTEP (CC No. 09-102 for indicator, 01-048A1 for bases), (AM-5762 for Indicator, AM-5719 for bases), cULus 2005/95/EC low voltage directive: EN60950-1:2001
12 in.
30.5cm
LCD
Gram, kilogram, ounce, pound, metric tones, custom
9.5cm
67
ABS Plastic, Stainless Steel, Aluminum