Learn More
OHAUS™ FD Food Portioning Scales
Speed, durability and cleanliness. OHAUS FD Food Portioning Scales are ideally suited for food preparation in restaurants, bakeries, food manufacturing and other areas requiring fast operation and flexibility.
Manufacturer:  OHAUS™ 83998147
 View more versions of this product
Catalog No. 02-113-28
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Standard Features
  • Rugged stainless-steel construction
  • Removable, dishwasher-safe, stainless-steel weighing pan
  • Simple 4-key operation and plain-text menu for fast setup
  • 3/4 in. high backlit LCD display with text annunciators Fast display update (under 2 seconds); large, dedicated tare key Auto tare for hands-free operation
  • Optional foot tare switch for hands-free operation
  • Checkweighing mode included with advanced features
  • 3-LED indication of Over, Accept and Under with audible buzzer
  • Positive mode for packaging, negative checkweighing mode for portioning, and “zero” checkweighing mode for sorting and grading operations
  • Dedicated Target key for entry of setpoints for checkweighing mode
Advanced Features
  • Selectable environmental averaging settings Backlight control with auto-off option Selectable weighing units Configurable checkweighing mode audible buzzer Configurable software lockout menu Span or linearity calibration in kg or lb. unit with optional external masses Selectable auto-zero tracking
  • NTEP - Legal for Trade certified
  • Low-battery detection system
  • Adjustable feet with integrated level bubble
Specifications
Specifications
0.0001kg
14 to 104°F
100/120V
3.6kg
9 in.
30cm
FD Food Portioning Balance
120 hr.
User selectable external span or linearity calibration
8.2 in.
20.9cm
LCD
Pound, decimal ounce, fractional ounce, gram, kilogram
71mm
43
Stainless Steel
<2 sec.
-10 to 40°C
8 lb.
8.2 in.
20.9cm
3 kg
4 Button membrane switch
Internal rechargeable Lead Acid battery
ISO 9001:2008, NSF/ANSI 169-2005, USDA/AMS, HACCP, cULus, AS/NZS4251.1, Emission, AS/NZS4252.1, Immunity, FCC Class B Part 15 operation, NTEP Class 3 to NIST Handbook 44, OIML R76 Class 3
2.8 in.
7.1cm
100/120V 50/60Hz
2.8 in.
50/60Hz
Rubber pan supports, Stainless Steel cover
FD3H