Learn More
OHAUS™ Navigator™ NV Portable Balances
SCI_ED
With dual multi-functional touchless infrared sensors allowing hands-free operation.
Specifications
Material ABS Plastic, Stainless Steel Pan
View More Specs
Products 1
Catalog Number Mfr. No. Model Capacity Readability Stabilization Time Price Quantity & Availability  
Catalog Number Mfr. No. Model Capacity Readability Stabilization Time Price Quantity & Availability  
S04792
SCI_ED
View Documents Promotion Details
Ohaus™
83033083/EMD
NV511 510 g 0.1 g 1 sec. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • With EC type approved models
  • Ultra-fast response time
  • Overload protection for wide variety of weighing applications
  • ABS plastic construction with stainless-steel pan
  • LCD with backlight display
  • Come with front level indicator (lit during power up), non-slip adjustable feet, stability indicator, external menu lock switch, shipping transportation lock, integrated security loop, and superior overload protection
  • Six full segment LCD white auto backlight with multiple icons and indicators
  • One second stabilization time
Applications
Weighing (14 units), Percentage weighing, Checkweighing, Parts counting, Accumulation/Totalization, and Display hold
Order Info
Units feature mains or battery operation with auto shut-off. Easy to install USB, RS232, and Ethernet interface options.
Certifications
UL listed
Specifications
Specifications
ABS Plastic, Stainless Steel Pan